Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046035.hg.1 |
TC02002851.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046035.hg.1 |
ENST00000295190.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046035.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046030.hg.1 |
TC02002851.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046030.hg.1 |
ENST00000295190.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046030.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046031.hg.1 |
TC02002851.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046031.hg.1 |
ENST00000295190.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046031.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046037.hg.1 |
TC02002851.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046037.hg.1 |
ENST00000295190.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02046037.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217498.hg.1 |
TC0200015971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217496.hg.1 |
TC0200015971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217490.hg.1 |
TC0200015971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217491.hg.1 |
TC0200015971.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000295190.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000412034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000425822.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000436869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
ENST00000457406.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015971.hg.1 |
NM_152527.4 |
Refseq |