Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000458582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000458582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000483911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000483911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000489887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
ENST00000489887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020329.hg.1 |
TC02001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020329.hg.1 |
ENST00000258411.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020329.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020328.hg.1 |
TC02001314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020328.hg.1 |
ENST00000258411.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02020328.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177182.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177182.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177182.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177182.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177182.hg.1 |
TC0200010818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200177181.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177181.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177181.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177181.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177181.hg.1 |
TC0200010818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
ENST00000458582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
ENST00000458582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200177183.hg.1 |
TC0200010818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000258411.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000458582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000458582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000483911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000483911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000489887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
ENST00000489887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010818.hg.1 |
NM_025216.2 |
Refseq |