Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017028445.1 | XP_016883934.1 | 7 | 2822 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |