Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
TC09000553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
ENST00000189978.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
ENST00000374447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
ENST00000374448.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
ENST00000416899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
ENST00000543335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
ENST00000545907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
NM_001166280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
NM_001166281.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006185.hg.1 |
NM_005592.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000189978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000374440.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000374448.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000416899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
NM_001166280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
NM_001166281.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
NM_005592.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000189978.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000374440.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000374448.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
ENST00000416899.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
NM_001166280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
NM_001166281.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
NM_005592.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156288.hg.1 |
TC0900008415.hg.1 |
Clariom D TC |