Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008735.hg.1 |
TC0X000661.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008735.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008735.hg.1 |
ENST00000415377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008735.hg.1 |
ENST00000535601.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008735.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X008735.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158092.hg.1 |
TC0X00008533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000218364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000425695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000448450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
ENST00000535601.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_001163280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008533.hg.1 |
NM_014500.4 |
Refseq |