Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004399.hg.1 |
TC08000305.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004399.hg.1 |
ENST00000518237.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004399.hg.1 |
ENST00000522495.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004399.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800150953.hg.1 |
TC0800007382.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000253513.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518237.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000518804.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000519154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000521636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000522840.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
ENST00000523779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007382.hg.1 |
NM_002164.5 |
Refseq |