Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
ENST00000262520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
ENST00000297679.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
ENST00000353250.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16005323.hg.1 |
TC16000365.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600154893.hg.1 |
TC1600007505.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000262520.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000297679.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000562932.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
ENST00000574447.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142777.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_001142778.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007505.hg.1 |
NM_025193.3 |
Refseq |