Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021514.hg.1 |
TC08001656.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021514.hg.1 |
ENST00000356838.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021514.hg.1 |
ENST00000377901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021514.hg.1 |
ENST00000523829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021514.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08021514.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000517538.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519187.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000519304.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522334.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000522594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012470.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
ENST00000356838.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
ENST00000377901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
ENST00000523829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
NM_001145153.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
NM_144649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800188206.hg.1 |
TC0800012470.hg.1 |
Clariom D TC |