Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023280.hg.1 |
ENST00000322886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023280.hg.1 |
ENST00000416496.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023280.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023280.hg.1 |
TC12001756.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023279.hg.1 |
ENST00000322886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023279.hg.1 |
ENST00000339887.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023279.hg.1 |
ENST00000416496.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023279.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023279.hg.1 |
TC12001756.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000547316.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000551558.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
ENST00000552907.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011330.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
ENST00000322886.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
ENST00000416496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
ENST00000550669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
NM_203394.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200187894.hg.1 |
TC1200011330.hg.1 |
Clariom D TC |