Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
ENST00000279036.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
ENST00000372689.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
ENST00000535404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
ENST00000543458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
NM_001184728.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
NM_001184729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
NM_015937.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005009.hg.1 |
TC20000347.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
ENST00000279035.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
ENST00000279036.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
ENST00000372689.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
ENST00000543458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
NM_001184728.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
NM_001184729.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
NM_001184730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
NM_015937.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20005016.hg.1 |
TC20000347.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000279036.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000372689.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000455050.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000543458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NM_001184728.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NM_001184729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NM_015937.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NR_047691.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NR_047692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NR_047693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
TC2000007488.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000279036.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000372689.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000455050.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
ENST00000543458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NM_001184728.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NM_001184729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NM_015937.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NR_047691.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NR_047692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152133.hg.1 |
NR_047693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000279035.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000279036.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000372689.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000455050.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000543458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_001184728.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_001184729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_001184730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_015937.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NR_047691.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NR_047692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NR_047694.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
TC2000007488.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000279035.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000279036.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000372689.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000455050.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
ENST00000543458.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_001184728.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_001184729.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_001184730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NM_015937.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NR_047691.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NR_047692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000152136.hg.1 |
NR_047694.1 |
Refseq |