Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008020.hg.1 |
ENST00000381621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008020.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008020.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008020.hg.1 |
TC20000553.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
ENST00000453770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009969.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
NM_178460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
TC2000009969.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
ENST00000381621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
ENST00000381623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
ENST00000429387.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170868.hg.1 |
NM_178460.2 |
Refseq |