Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008466.hg.1 |
ENST00000202578.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008466.hg.1 |
ENST00000344754.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008466.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008466.hg.1 |
TC20000575.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158073.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158073.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158073.hg.1 |
TC2000008228.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158073.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158073.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000344754.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
ENST00000419548.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008228.hg.1 |
NM_023068.3 |
Refseq |