Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017011318.1 | XP_016866807.1 | 1 - 2 | 1382 | 69 | |
GenBank mRNA | AK074110.1 | - | 5 - 6 | 2313 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |