Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012044.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012044.hg.1 |
ENST00000357199.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012044.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012044.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012044.hg.1 |
TC20000882.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012043.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012043.hg.1 |
ENST00000357199.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012043.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012043.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012043.hg.1 |
TC20000882.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012045.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012045.hg.1 |
ENST00000357199.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012045.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012045.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20012045.hg.1 |
TC20000882.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
TC2000009231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164578.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
TC2000009231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164579.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009231.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
NM_181510.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
TC2000009231.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
ENST00000289953.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
ENST00000357199.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
NM_130896.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164580.hg.1 |
NM_181510.2 |
Refseq |