Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
TC01002945.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
ENST00000370031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
ENST00000370032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
ENST00000402983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045764.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045767.hg.1 |
TC01002945.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045767.hg.1 |
ENST00000370031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045767.hg.1 |
ENST00000370032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045767.hg.1 |
ENST00000402983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045767.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045767.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
TC01002945.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
ENST00000370031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
ENST00000370032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
ENST00000402983.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01045766.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212997.hg.1 |
TC0100015171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100213000.hg.1 |
TC0100015171.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000483729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015171.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000370031.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000370032.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000402983.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000420055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
ENST00000493676.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
NM_001102592.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
NM_144584.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100212999.hg.1 |
TC0100015171.hg.1 |
Clariom D TC |