Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016657.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016657.hg.1 |
ENST00000543625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016657.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016657.hg.1 |
TC11001309.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016658.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016658.hg.1 |
ENST00000543625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016658.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016658.hg.1 |
TC11001309.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016659.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016659.hg.1 |
ENST00000543625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016659.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016659.hg.1 |
TC11001309.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100174729.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174729.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174729.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174729.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174729.hg.1 |
TC1100009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174731.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174731.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174731.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174731.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174731.hg.1 |
TC1100009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174728.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174728.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174728.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174728.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174728.hg.1 |
TC1100009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174730.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174730.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174730.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174730.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174730.hg.1 |
TC1100009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000254436.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
ENST00000533692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009880.hg.1 |
NM_003141.3 |
Refseq |