Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019247.hg.1 |
TC08001480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019247.hg.1 |
ENST00000311812.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019247.hg.1 |
ENST00000428383.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08019247.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800176746.hg.1 |
TC0800011260.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000311812.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000428383.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000518342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000519521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520352.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520661.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
ENST00000520743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011260.hg.1 |
NM_152628.3 |
Refseq |