Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016042.hg.1 |
TC06001337.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016042.hg.1 |
ENST00000244527.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016042.hg.1 |
ENST00000427328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016042.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016042.hg.1 |
ENST00000476801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016042.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016043.hg.1 |
TC06001337.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016043.hg.1 |
ENST00000244527.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016043.hg.1 |
ENST00000427328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016043.hg.1 |
ENST00000476801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016043.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016041.hg.1 |
TC06001337.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016041.hg.1 |
ENST00000244527.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016041.hg.1 |
ENST00000427328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016041.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016041.hg.1 |
ENST00000476801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016041.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016046.hg.1 |
TC06001337.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016046.hg.1 |
ENST00000244527.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016046.hg.1 |
ENST00000427328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016046.hg.1 |
ENST00000476801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016046.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175156.hg.1 |
TC0600011112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175154.hg.1 |
TC0600011112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011112.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175159.hg.1 |
TC0600011112.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000244527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000377886.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000468082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
ENST00000476801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
NM_005074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600175155.hg.1 |
TC0600011112.hg.1 |
Clariom D TC |