Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017214.hg.1 |
TC0X001270.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017214.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017214.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010493.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171542.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171542.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171542.hg.1 |
ENST00000218006.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171542.hg.1 |
NM_001522.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171542.hg.1 |
TC0X00010493.hg.1 |
Clariom D TC |