Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001617.hg.1 |
ENST00000162391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001617.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001617.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001617.hg.1 |
TC12000118.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000428177.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006700.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
ENST00000162391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
ENST00000539192.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
NM_018416.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200146859.hg.1 |
TC1200006700.hg.1 |
Clariom D TC |