Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040875.2 | NP_001035808.1 | 2 - 3 | 262 | 121 | |
NM_018972.2 | NP_061845.2 | 2 - 3 | 392 | 121 | ||
XM_017013585.1 | XP_016869074.1 | 2 - 3 | 1524 | 121 | ||
XM_017013586.1 | XP_016869075.1 | 2 - 3 | 1524 | 121 | ||
GenBank mRNA | AB551556.1 | - | 2 - 3 | 391 | 121 | |
AB551557.1 | - | 2 - 3 | 391 | 121 | ||
AB551558.1 | - | 2 - 3 | 389 | 121 | ||
AK292572.1 | - | 2 - 3 | 331 | 121 | ||
BC024939.1 | - | 2 - 3 | 227 | 3923335 | 121 | |
BC035450.1 | - | 2 - 3 | 239 | 121 | ||
DB505741.1 | - | 2 - 3 | 392 | 121 | ||
Y17849.1 | - | 2 - 3 | 320 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |