Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001107.hg.1 |
ENST00000329493.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001107.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001107.hg.1 |
TC19000071.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001106.hg.1 |
ENST00000329493.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001106.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001106.hg.1 |
TC19000071.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900146441.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146441.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146441.hg.1 |
TC1900006642.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146441.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146441.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146442.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146442.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146442.hg.1 |
TC1900006642.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146442.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146442.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
ENST00000329493.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006642.hg.1 |
NM_001135580.1 |
Refseq |