Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013952.hg.1 |
ENST00000320152.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013952.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10013952.hg.1 |
TC10001110.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000464584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
ENST00000464584.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168605.hg.1 |
TC1000010070.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000320152.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376376.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000376378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000464584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
ENST00000464584.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_001282786.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010070.hg.1 |
NM_020200.6 |
Refseq |