Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034542.hg.1 |
TC01002263.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034542.hg.1 |
ENST00000335496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034542.hg.1 |
ENST00000466212.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034542.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034542.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196145.hg.1 |
TC0100013042.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000335496.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000375577.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000466212.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000491418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000540400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
ENST00000612240.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013042.hg.1 |
NM_198546.1 |
Refseq |