Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015797.hg.1 |
TC03000897.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015797.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000486295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000486295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000486295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000510136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000510136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000510136.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167714.hg.1 |
TC0300009422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000382864.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000431685.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000461151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000482245.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000483846.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000486295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000486295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000486295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000488647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000491953.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502332.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000502778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000506760.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000510136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000510136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000510136.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
ENST00000511230.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009422.hg.1 |
NR_021485.2 |
Refseq |