Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012733.hg.1 |
TC05000872.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012733.hg.1 |
ENST00000231198.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012733.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012730.hg.1 |
TC05000872.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012730.hg.1 |
ENST00000231198.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012730.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012734.hg.1 |
TC05000872.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012734.hg.1 |
ENST00000231198.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012734.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164594.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164594.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164594.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164594.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164594.hg.1 |
TC0500009223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164598.hg.1 |
TC0500009223.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
ENST00000523575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009223.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
ENST00000231198.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
ENST00000521655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
NM_017872.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164599.hg.1 |
TC0500009223.hg.1 |
Clariom D TC |