Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045944.hg.1 |
TC02002848.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045944.hg.1 |
ENST00000283943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045944.hg.1 |
ENST00000389044.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045944.hg.1 |
ENST00000389045.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045944.hg.1 |
NM_004238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045945.hg.1 |
TC02002848.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045945.hg.1 |
ENST00000283943.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045945.hg.1 |
ENST00000389044.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045945.hg.1 |
ENST00000389045.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02045945.hg.1 |
NM_004238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
ENST00000283943.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
ENST00000389044.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
ENST00000389045.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_001284214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_001284215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_001284216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_004238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
ENST00000283943.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
ENST00000389044.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
ENST00000389045.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_001284214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_001284215.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_001284216.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
NM_004238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200217394.hg.1 |
TC0200015963.hg.1 |
Clariom D TC |