Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007939.hg.1 |
ENST00000371174.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007939.hg.1 |
ENST00000419175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007939.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007939.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10007939.hg.1 |
TC10000684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008539.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
ENST00000371174.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
ENST00000630152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
NM_001017520.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
NM_004088.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000158255.hg.1 |
TC1000008539.hg.1 |
Clariom D TC |