Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057872.hg.1 |
TC01003777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057872.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057872.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229953.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229953.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229953.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100229953.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100229953.hg.1 |
TC0100017142.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000310833.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000356522.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000367036.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
ENST00000485761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001025109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017142.hg.1 |
NM_001773.2 |
Refseq |