Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
TC05003407.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000265334.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000435211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000435240.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000518409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000521118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000523054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000523832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
ENST00000536186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
NM_001113575.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024357.hg.1 |
NM_016508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000265334.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000520592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000520693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000521118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000523054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000523832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
NM_001113575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
NM_001300853.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
NM_016508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000265334.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000520592.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000520693.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000521118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000523054.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
ENST00000523832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
NM_001113575.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
NM_001300853.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
NM_016508.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500196673.hg.1 |
TC0500013360.hg.1 |
Clariom D TC |