Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000293761.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000570836.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000572265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000572265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000572265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000572265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000572265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000573740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000573740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000573740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000573740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000573740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000573740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000574640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576394.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
ENST00000576572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009520.hg.1 |
NM_001140.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007970.hg.1 |
ENST00000340858.4 |
Ensembl |