Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011880.hg.1 |
TC05000829.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011880.hg.1 |
ENST00000230671.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011880.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011880.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000513403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009085.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
ENST00000230671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
ENST00000524041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
NM_014228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500163430.hg.1 |
TC0500009085.hg.1 |
Clariom D TC |