Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001288.hg.1 |
ENST00000333870.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001288.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001288.hg.1 |
TC17000095.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146996.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146996.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146996.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146996.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146996.hg.1 |
TC1700006746.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006746.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
ENST00000333870.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
ENST00000574034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
NM_175734.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700146997.hg.1 |
TC1700006746.hg.1 |
Clariom D TC |