Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303256.2 | NP_001290185.1 | 21 - 22 | 2770 | 112 | |
NM_001303257.2 | NP_001290186.1 | 21 - 22 | 2770 | 112 | ||
NM_014941.3 | NP_055756.1 | 22 - 23 | 3636 | 112 | ||
XM_011530004.1 | XP_011528306.1 | 21 - 22 | 2995 | 112 | ||
XM_017028667.1 | XP_016884156.1 | 21 - 22 | 2995 | 112 | ||
XM_017028668.1 | XP_016884157.1 | 22 - 23 | 3090 | 112 | ||
GenBank mRNA | AB020659.1 | - | 22 - 23 | 3550 | 112 | |
AL133637.1 | - | 3 - 4 | 241 | 112 | ||
BC019257.1 | - | 20 - 21 | 2297 | 3503651 | 112 | |
BC136782.1 | - | 22 - 23 | 2495 | 9020772 | 112 | |
CR456469.1 | - | 20 - 21 | 2296 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |