Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000295797.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000476635.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000482353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000482353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000482353.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000483697.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000483697.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000483697.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000483697.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000485837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000488541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000488541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000488541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000493761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000493761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000493761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
ENST00000493761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009468.hg.1 |
NM_002740.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010363.hg.1 |
ENST00000392722.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010363.hg.1 |
ENST00000392722.3 |
Ensembl |