Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011509661.1 | XP_011507963.1 | 13 - 14 | 1502 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |