Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000476018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000476018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
ENST00000476018.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000298454.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000476018.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000476018.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
ENST00000476018.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010589.hg.1 |
NM_144984.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172235.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172235.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172235.hg.1 |
ENST00000332853.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000172235.hg.1 |
NM_001031746.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000172235.hg.1 |
TC1000010589.hg.1 |
Clariom D TC |