Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
225547_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016580.hg.1 |
TC08001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016580.hg.1 |
ENST00000502076.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016580.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016581.hg.1 |
TC08001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016581.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016577.hg.1 |
TC08001290.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016577.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000364849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
ENST00000522478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172573.hg.1 |
TC0800010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172575.hg.1 |
TC0800010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172570.hg.1 |
TC0800010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172574.hg.1 |
TC0800010695.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000364849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520619.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000520944.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521127.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000521399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
ENST00000522478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010695.hg.1 |
NR_002599.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
225547_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
225547_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NR_002599_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
225547_at5 |
- |
- |
Human Genome U95 Set |
48853_PM_at |
- |
- |
Human Genome U95 Set |
48853_at |
- |
- |
Human Genome U133 Set |
225547_at |
- |
- |
Human Genome U133 Set |
225547_PM_at |
- |
- |