Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022936.hg.1 |
TC01001479.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022936.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022936.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022938.hg.1 |
TC01001479.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022938.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022938.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022938.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022941.hg.1 |
TC01001479.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022941.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022941.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022941.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177717.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177717.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177717.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177717.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177717.hg.1 |
TC0100010559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010559.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177721.hg.1 |
TC0100010559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
ENST00000367764.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177719.hg.1 |
TC0100010559.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177716.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177716.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177716.hg.1 |
ENST00000439373.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177716.hg.1 |
NM_001136107.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177716.hg.1 |
TC0100010559.hg.1 |
Clariom D TC |