Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010328.hg.1 |
ENST00000298472.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010328.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010328.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010328.hg.1 |
TC10000850.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161797.hg.1 |
TC1000009017.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000298472.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
ENST00000497497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009017.hg.1 |
NM_003054.4 |
Refseq |