Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026014.hg.1 |
TC03001442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026014.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026014.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026013.hg.1 |
TC03001442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026013.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026013.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026015.hg.1 |
TC03001442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026015.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03026015.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183072.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183072.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183072.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183072.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183072.hg.1 |
TC0300011146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300183073.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183073.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183073.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300183073.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300183073.hg.1 |
TC0300011146.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
ENST00000232888.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011146.hg.1 |
NM_004704.4 |
Refseq |