Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000489.4 | NP_000480.3 | 28 - 29 | 6599 | 143 | |
NM_138270.3 | NP_612114.2 | 27 - 28 | 6485 | 143 | ||
XM_005262153.4 | XP_005262210.2 | 28 - 29 | 6596 | 143 | ||
XM_005262154.4 | XP_005262211.2 | 29 - 30 | 6512 | 143 | ||
XM_005262155.4 | XP_005262212.2 | 28 - 29 | 6700 | 143 | ||
XM_005262156.4 | XP_005262213.2 | 27 - 28 | 6452 | 143 | ||
XM_005262157.4 | XP_005262214.2 | 28 - 29 | 6395 | 143 | ||
XM_006724666.3 | XP_006724729.1 | 27 - 28 | 6482 | 143 | ||
XM_006724667.3 | XP_006724730.1 | 26 - 27 | 6338 | 143 | ||
XM_017029601.1 | XP_016885090.1 | 29 - 30 | 6509 | 143 | ||
XM_017029602.1 | XP_016885091.1 | 28 - 29 | 6680 | 143 | ||
XM_017029603.1 | XP_016885092.1 | 27 - 28 | 6449 | 143 | ||
XM_017029604.1 | XP_016885093.1 | 28 - 29 | 6398 | 143 | ||
XM_017029605.1 | XP_016885094.1 | 29 - 30 | 6613 | 143 | ||
XM_017029606.1 | XP_016885095.1 | 27 - 28 | 6586 | 143 | ||
XM_017029607.1 | XP_016885096.1 | 27 - 28 | 6583 | 143 | ||
XM_017029608.1 | XP_016885097.1 | 26 - 27 | 6335 | 143 | ||
XM_017029609.1 | XP_016885098.1 | 28 - 29 | 6499 | 143 | ||
XM_017029610.1 | XP_016885099.1 | 28 - 29 | 6496 | 143 | ||
XM_017029611.1 | XP_016885100.1 | 27 - 28 | 6251 | 143 | ||
GenBank mRNA | AB102641.1 | - | 28 - 29 | 6331 | 143 | |
AK295107.1 | - | 11 - 12 | 1451 | 143 | ||
AK296959.1 | - | 7 - 8 | 834 | 143 | ||
BX647222.1 | - | 3 - 4 | 3403 | 143 | ||
U09820.1 | - | 20 - 21 | 3810 | 143 | ||
U72936.2 | - | 29 - 30 | 6668 | 143 | ||
U72937.2 | - | 28 - 29 | 6546 | 143 | ||
U72938.2 | - | 27 - 28 | 6432 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |