Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011598.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011598.hg.1 |
ENST00000540413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011598.hg.1 |
ENST00000542049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011598.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011598.hg.1 |
TC19000866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011599.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011599.hg.1 |
ENST00000540413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011599.hg.1 |
ENST00000542049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011599.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011599.hg.1 |
TC19000866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011600.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011600.hg.1 |
ENST00000540413.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011600.hg.1 |
ENST00000542049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011600.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19011600.hg.1 |
TC19000866.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168689.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168689.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168689.hg.1 |
TC1900008825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900168689.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168689.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168691.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168691.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168691.hg.1 |
TC1900008825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900168691.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168691.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000419486.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000474397.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
ENST00000479081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008825.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168690.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168690.hg.1 |
NM_002739.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168690.hg.1 |
TC1900008825.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900168690.hg.1 |
ENST00000263431.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168690.hg.1 |
NM_002739.4 |
Refseq |