Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014786.3 | NP_055601.2 | 10 - 11 | 4226 | 66 | |
XM_017018623.1 | XP_016874112.1 | 10 - 11 | 2617 | 66 | ||
XM_017018624.1 | XP_016874113.1 | 9 - 10 | 1696 | 66 | ||
GenBank mRNA | AB002335.1 | - | 10 - 11 | 3000 | 66 | |
AF378754.1 | - | 10 - 11 | 4220 | 66 | ||
BC137228.1 | - | 10 - 11 | 4226 | 9021225 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |