Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005072.hg.1 |
TC01000332.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005072.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005072.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005072.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100151828.hg.1 |
TC0100007434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000374280.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000470037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000481377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
ENST00000484339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007434.hg.1 |
NM_004455.2 |
Refseq |