Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024528.3 | NP_078804.2 | 2 - 3 | 629 | 100 | |
XM_017029842.1 | XP_016885331.1 | 2 - 3 | 166 | 100 | ||
GenBank mRNA | AK026279.1 | - | 2 - 3 | 629 | 100 | |
AL832118.1 | - | 4 - 5 | 570 | 100 | ||
AY388958.1 | - | 2 - 3 | 629 | 100 | ||
BC000940.2 | - | 2 - 3 | 510 | 100 | ||
BC015354.1 | - | 2 - 3 | 493 | 3450847 | 100 | |
BC016783.1 | - | 2 - 3 | 497 | 100 | ||
BC018968.1 | - | 2 - 3 | 600 | 100 | ||
BC032442.1 | - | 2 - 3 | 487 | 5139142 | 100 | |
BC071686.1 | - | 2 - 3 | 608 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons |