Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014727.2 | NP_055542.1 | 22 - 23 | 4918 | 76 | |
XM_011527561.2 | XP_011525863.2 | 22 - 23 | 4789 | 76 | ||
XM_011527562.2 | XP_011525864.1 | 22 - 23 | 5119 | 76 | ||
XM_017027544.1 | XP_016883033.1 | 22 - 23 | 5119 | 76 | ||
XM_017027545.1 | XP_016883034.1 | 21 - 22 | 4832 | 76 | ||
XM_017027546.1 | XP_016883035.1 | 17 - 18 | 2040 | 76 | ||
GenBank mRNA | AB002302.3 | - | 20 - 21 | 4018 | 76 | |
AF104918.1 | - | 20 - 21 | 2932 | 76 | ||
AF186605.1 | - | 22 - 23 | 4590 | 76 | ||
AJ007041.1 | - | 22 - 23 | 4918 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons |