Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000262630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000392197.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000426659.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000442282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000451726.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
ENST00000481182.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007876.hg.1 |
NM_014383.2 |
Refseq |