Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000430852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000430852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
ENST00000380092.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
ENST00000380094.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012513.hg.1 |
TC10001004.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000430852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000430852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009685.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000191063.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000380092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000380094.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
ENST00000492368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
NM_001009941.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
NM_001009943.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
NM_019046.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166170.hg.1 |
TC1000009685.hg.1 |
Clariom D TC |