Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301060.1 | NP_001287989.1 | 3 - 4 | 463 | 81 | |
NM_024848.2 | NP_079124.1 | 3 - 4 | 463 | 81 | ||
NR_125361.1 | 2 - 3 | 391 | 81 | |||
XM_005244798.2 | XP_005244855.1 | 2 - 3 | 327 | 81 | ||
XM_011542169.1 | XP_011540471.1 | 3 - 4 | 463 | 81 | ||
XM_011542170.2 | XP_011540472.1 | 3 - 4 | 463 | 81 | ||
XM_011542172.2 | XP_011540474.1 | 2 - 3 | 327 | 81 | ||
XM_011542173.2 | XP_011540475.1 | 3 - 4 | 463 | 81 | ||
XM_011542174.2 | XP_011540476.1 | 3 - 4 | 463 | 81 | ||
XM_011542176.1 | XP_011540478.1 | 3 - 4 | 463 | 81 | ||
XM_011542177.1 | XP_011540479.1 | 3 - 4 | 463 | 81 | ||
XM_017002365.1 | XP_016857854.1 | 3 - 4 | 463 | 81 | ||
GenBank mRNA | AK024003.1 | - | 3 - 4 | 266 | 81 | |
AK299217.1 | - | 2 - 3 | 311 | 81 | ||
AK310323.1 | - | 2 - 3 | 327 | 81 | ||
BC021704.2 | - | 3 - 4 | 419 | 4825983 | 81 | |
DB041436.1 | - | 3 - 4 | 463 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |